Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Reagents for cobas c 111 Chemistry Analyzer

Catalog No. 22045891 Shop All Roche Diagnostics Products
Change view
Click to view available options
Quantity:
100 Tests
1000 mL
200 Tests
400 Tests
6.3 mL
Type:
Calcium Reagent
Creatine Kinase-MB Reagent
GGT-2 Reagent
HbA1c Hemolyzing Reagent
Hemolyzing Reagent Gen.2 Reagent
Iron Reagent
Lactate Reagent
Lipase Reagent Kit
Uric Acid Gen.2 Reagent
a-Amylase Reagent
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-045-891 100 Tests Lipase Reagent Kit
22-045-779 400 Tests Calcium Reagent
22-045-883 100 Tests Creatine Kinase-MB Reagent
22-045-888 200 Tests GGT-2 Reagent
22-045-797 400 Tests Uric Acid Gen.2 Reagent
22-045-853 6.3 mL Hemolyzing Reagent Gen.2 Reagent
22-045-854 1000 mL HbA1c Hemolyzing Reagent
22-045-895 100 Tests Iron Reagent
22-044-871 100 Tests Lactate Reagent
22-045-880 200 Tests a-Amylase Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 22-045-891 Supplier Roche Diagnostics Supplier No. 05401704190
This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.