Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Real Time Electrophoresis System, with Power Supply

Catalog No. 14955208 Shop All Fisherbrand Products
Change view
Click to view available options
Plug Type:
US
Universal
Voltage:
100/115 V
220/240 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Voltage
14-955-208 Universal 220/240 V
14-955-207 US 100/115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-955-208
Add to Cart
Add to Cart

Run DNA gels, view separations as they occur, and capture gel images using a smart phone

  • Tool for small DNA gel runs
  • Power supply has selectable voltage of 100 V, 50 V, or 35 V and timer
  • Compatible with SYBR, Gel green, SmartGlow, and other green fluorescent DNA stains
  • Imaging enclosure and photo filter allows gel imaging with any smart phone camera
  • Includes:
    • Gel running tank
    • Direct connect power supply
    • Amber filter lid
    • Small blue LED transilluminator
    • Gel casting equipment (for casting your own gels)
      • Stand
      • Trays
      • Combs

Order Info

Voltage: 220/240 V

Specifications

Buffer 0.5 x or 1 x TAE or TBE
Certifications/Compliance CE
Color Gray and clear
Detection Method Smart phone
For Use With (Equipment) Standard Electrophoresis Power Supply
Gel Compatibility Make gels using included casting set
Type DNA
Display Digital LED timer
Gel Dimensions 10.5 x 10 cm, 10.5 x 6 cm
Gel Percentage 0.5 to 6%
Gel Type Agarose
Green Features Uses minimal consumables, no waste associated with precast gels
Includes Gel tank, Blue LED illuminator, Filter cover, Gel casting set, Smart phone imaging enclosure, 230 to 115 V transformer
Label or Dye Use with SmartGlow or SYBR green stain
Material Molded polycarbonate
Mode of Separation Electrophoresis
No. of Gels 1
No. of Wells 12 or 22
No. per Pack 1 Ea.
Plug Type Universal
Sample Loading Volume 10 to 20 μL
Sample Type Nucleic acid
Teeth Count 12 or 22
Thickness (Metric) Teeth 1 mm, 2 mm
Volume (Metric) Buffer 230 mL
Volume (Metric) Chamber 230 mL
Volume (Metric) Sample 10 to 20 μL
Volume (Metric) Well 10 to 20 μL
Warranty 2 years
Dimensions (L x W) 7.5 x 8.25 in.
Dimensions (L x W x H) 7.5 x 8.25 x 5 in.
Length (English) 7.5 in.
Length (English) Footprint 7.5 in.
Length (Metric) 8.25 in.
Length (Metric) Footprint 21 in.
Length (Metric) Gel 10 cm
Width (Metric) Gel 10.5 cm
Width (Metric) Teeth 3 mm, 5.65 mm
Width (Metric) Well 3 mm, 5.65 mm
Height (English) Footprint 5 in.
Height (Metric) Footprint 14 cm
Weight (English) 1.65 lb.
Weight (Metric) 0.75 kg
Voltage 220/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.