Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Real Time Electrophoresis Tank System

Catalog No. 14955206 Shop All Fisherbrand Products
Change view
Click to view available options
Voltage:
100/115 V
220/240 V
Plug Type:
US
Universal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage Plug Type
14-955-206 220/240 V Universal
14-955-205 100/115 V US
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-955-206
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Run DNA gels, view separations as they occur, and capture gel images using a smart phone

  • Tool for small DNA gel runs
  • Connects to any standard electrophoresis power supply
  • Compatible with SYBR, Gel green, SmartGlow, and other green fluorescent DNA stains
  • Imaging enclosure and photo filter allo gel imaging with any smart phone camera
  • Includes:
    • Gel running tank
    • Amber filter lid
    • Small blue LED transilluminator
    • Gel casting equipment (for casting your own gels)
      • Stand
      • Trays
      • Combs
  • Specifications

    Buffer 0.5 x or 1 x TAE or TBE
    Certifications/Compliance CE
    Detection Method Smart phone
    For Use With (Equipment) Standard Electrophoresis Power Supply
    Gel Compatibility Make gels using included casting set
    Type DNA
    Gel Dimensions 10.5 x 10 cm, 10.5 x 6 cm
    Gel Percentage 0.5 to 6%
    Gel Type Agarose
    Green Features Uses minimal consumables, no waste associated with precast gels
    Includes Gel tank, Blue LED illuminator, Filter cover, Gel casting set, Smart phone imaging enclosure
    Label or Dye Use with SmartGlow or SYBR Green Stain
    Material Molded polycarbonate
    Mode of Separation Electrophoresis
    No. of Gels 1
    No. of Wells 12 or 22
    No. per Pack 1 Ea.
    Plug Type Universal
    Power Requirements 120 V
    Sample Loading Volume 10 to 20 μL
    Sample Type Nucleic acid
    Teeth Count 12 or 22
    Thickness (Metric) Teeth 1 mm, 2 mm
    Volume (Metric) Buffer 230 mL
    Volume (Metric) Chamber 230 mL
    Volume (Metric) Sample 10 to 20 μL
    Volume (Metric) Well 10 to 20 μL
    Warranty 2 years
    Dimensions (L x W) 7.5 x 8.25 in.
    Dimensions (L x W x H) 7.5 x 8.25 x 5 in.
    Length (English) 7.5 in.
    Length (English) Footprint 7.5 in.
    Length (Metric) 8.25 in.
    Length (Metric) Footprint 21 in.
    Length (Metric) Gel 10 cm
    Width (Metric) Gel 10.5 cm
    Width (Metric) Teeth 3 mm, 5.65 mm
    Width (Metric) Well 3 mm, 5.65 mm
    Height (English) Footprint 5 in.
    Height (Metric) 0.5 cm
    Height (Metric) Footprint 14 cm
    Weight (English) 1.65 lb.
    Weight (Metric) 0.75 kg
    Electrical Requirements 120 V
    Voltage 220/240 V
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.