Learn More
Bachem rec Brain-Derived Neurotrophic Factor (human)
SDP
Supplier:  Bachem 4038290.0010
Encompass_Preferred
rec Brain-Derived Neurotrophic Factor (human), 10ug. rec BDNF (human). Brain-derived neurotrophic factor (BDNF) is a member of the neurotrophin family. Its expression in the human central nervous system is altered in neurodegenerative diseases like Alzheimer's and Parkinson's disease. BDNF promotes survival of dopaminergic neurons and protects against Ab-induced neurotoxicity in vitro and in vivo.
Catalog No. 50-383-090
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.