Learn More
Biotang Inc Desmopressin, DDAVP, Desmopressin also called ADH (Anti-Diuretic Hormone) has a molecular formula of C46H64N14O12S2, Mpr-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly-NH2 having a Mw of 1069.23
Supplier:  Biotang Inc BTHOR270
Encompass_Preferred
Desmopressin, DDAVP, Desmopressin also called ADH (Anti-Diuretic Hormone) has a molecular formula of C46H64N14O12S2, Mpr-Tyr-Phe-Gln-Asn-Cys-Pro-D-Arg-Gly-NH2 having a Mw of 1069.23
Catalog No. 50-148-978
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.