Learn More
Abcam Recombinant Human KRAS (mutated G12D) protein (Tagged-His Tag), 20UG
Supplier:  Abcam AB26871220UG
Encompass_Preferred
Recombinant Human KRAS (mutated G12D) protein (Tagged-His Tag) is a Human Full Length protein, in the 2 to 186 aa range, expressed in Escherichia coli, with >85% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: KRAS2, RASK2, KRAS, GTPase KRas, K-Ras 2, Ki-Ras, c-K-ras, c-Ki-ras
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2010487