Learn More
Abcam Recombinant Human Lysozyme protein, 10UG
Supplier:  Abcam AB15883910UG
Encompass_Preferred
Recombinant Human Lysozyme protein is a Human Fragment protein, in the 19 to 118 aa range, expressed in Wheat germ and suitable for ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: LZM, LYZ, Lysozyme C
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2010485
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.