Learn More
Biotang Inc Ku P70/P80 Human Recombinant, produced in SF9 insect cells is a glycosylated, polypeptide chain having a molecular mass of 70,638 Dalton for the p70 subunit and 83,528 Dalton for the p80 subunit. Ku (P70/P80) is expressed with a -6xHis tag
Supplier:  Biotang Inc BTPRO397
Encompass_Preferred
Ku P70/P80 Human Recombinant, produced in SF9 insect cells is a glycosylated, polypeptide chain having a molecular mass of 70,638 Dalton for the p70 subunit and 83,528 Dalton for the p80 subunit. Ku (P70/P80) is expressed with a -6xHis tag
Catalog No. 50-149-954
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.