Learn More
ILEX LIFE SCIENCES LLC REC HUMAN ALBUMIN POWDER 5X1G
SDP
Supplier:  ILEX LIFE SCIENCES LLC PB020001GX05
Encompass_Preferred
Provis Biolabs Recombinant Human Albumin (rHA), Lyophilized Powder, GMP Grade (5 x 1 g). DESCRIPTION: Recombinant Human Albumin (rHA), GMP Grade, expressed in Pichia pastoris, is a 66 kDa synthetic alternative to human serum albumin (HSA). The rHA is a highly purified product that is free of animal or human components. It is developed as an alternative to HSA, to which it is structurally equivalent. The rHA offers a reliable and sustainable alternative to traditional HSA sources. Purity levels ≥ 99% confirmed by orthogonal quality control tests. Raw materials used are animal component free, TSE/BSE-free certified, and comply with IP/USP/EP. APPLICATIONS: Cell and tissue culture media supplement, used as serum substitute in serum-free xeno-free cell culture, medium component for cryopreservation and stem cell culture, stabilizer protein for pharmaceutical and biotech products, reduces unspecific binding in molecular biology assays.
Catalog No. 50-273-3642
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.