Learn More
Biotang Inc Prolactin Rat Recombinant, Mammotropin, Luterotropic hormone, Lutetropin, PRL. in E.Coli is a single, non-glycosylated polypeptide chain containing 198 amino acids and having a molecular mass of 22.6 kDa.
Supplier:  Biotang Inc BTCYT322
Encompass_Preferred
Prolactin Rat Recombinant, Mammotropin, Luterotropic hormone, Lutetropin, PRL. in E.Coli is a single, non-glycosylated polypeptide chain containing 198 amino acids and having a molecular mass of 22.6 kDa.
Catalog No. 50-151-038
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.