Learn More
Biotang Inc Streptavidin Recombinant, His Tag, produced in E.Coli is a single, non-glycosylated polypeptide chain containing 167 amino acids and having a molecular mass of 17 kDa.
Supplier:  Biotang Inc BTPRO621
Encompass_Preferred
Streptavidin Recombinant, His Tag, produced in E.Coli is a single, non-glycosylated polypeptide chain containing 167 amino acids and having a molecular mass of 17 kDa.
Catalog No. 50-151-073