Learn More
New England Biolabs, Inc. RecA – 1000 µg
SDP
Supplier:  New England Biolabs, Inc. M0249L
Encompass
E. coli RecA is necessary for genetic recombination, reactions involving DNA repair and UV-induced mutagenesis. RecA promotes the autodigestion of the lexA repressor, umuD protein and lambda repressor. Cleavage of LexA derepresses more than 20 genes. In vitro studies indicate that in the presence of ATP, RecA promotes the strand exchange of single-strand DNA fragments with homologous duplex DNA. The reaction has three distinct steps: (i) RecA polymerizes on the single-strand DNA, (ii) the nucleoprotein filament binds the duplex DNA and searches for a homologous region, (iii) the strands are exchanged.
Catalog No. 50-435-7
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.