Learn More
Abcam Recombinant Anti-PRDM1/Blimp1 antibody [6D3]Key features and details:Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supplyRat monoclonal [6D3] to PRDM1/Blimp1Suitable for: WB, IHC-PReacts with: Mouse100ug
Supplier:  Abcam AB243146100UG
Encompass_Preferred
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Recombinant Anti-PRDM1/Blimp1 antibody [6D3]Key features and details:Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supplyRat monoclonal [6D3] to PRDM1/Blimp1Suitable for: WB, IHC-PReacts with: Mouse100ug
Catalog No. NC2201862
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.