Learn More
Market Lab Labels for 3 Line Label Gun, 6000pk
Supplier:  Market Lab 6707WH
Encompass_Preferred
The MLI ML6707-WH is a 3-line label gun that prints white labels. It uses 1" x 3" labels and prints up to 3 lines of text at 14 characters per line. The label gun comes with a user guide and starter roll of 500 white labels. It uses inkless technology to print on the labels. The label gun can print labels for file folders, shelves, boxes, and more. It is lightweight and portable.
Catalog No. NC1831335
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.