Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Rechargeable LiteBox™ Lanterns

Catalog No. 19819830 Shop All Streamlight Products
Change view
Click to view available options
Color:
Orange
Yellow
Description:
Power-Failure System
Swivel neck, Standard System
Wattage:
8 W
20 W
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Description Wattage
19-819-830 Orange Swivel neck, Standard System 8 W
19-041-961 Yellow Swivel neck, Standard System 20 W
18-999-1651 Orange Power-Failure System 8 W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-819-830 Supplier Streamlight™ Supplier No. 45116

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Adjustable swivel neck allows for rotation of the lamp head

  • Light source: High-intensity halogen bi-pin bulb in 8 or 20w spot for 8 or 20w bulbs; 3.5 for 20w
  • Light output: see Ordering Table
  • 12AH battery provides up to nine hours of run time; battery is rechargeable up to 500 times
  • LEDs provide battery status: red is charging, green is fully charged
  • High-impact ABS thermoplastic construction sealed to resist water, sand and dust
  • High-intensity sealed halogen beam
  • Rubberized impact bumper and a protective lens ring

Standard System
Ideal for field maintenance, inspection of construction sites, and remote equipment; international safety orange or high-visibility yellow; includes shoulder strap; comes with AC and DC chargers and charge rack.

Vehicle Mount System
International safety orange or high-visibility yellow; comes with vehicle-mount hard-wire rack and shoulder carrying strap.

Power Failure System
International safety orange or beige; comes with AC/DC chargers and smart power failure circuitry that automatically goes on when the power goes off.

Order Info

Dimensions: L x W x H: 11.5 x 5.1 x 7 in. (29.2 x 13 x 17.8cm).

Certifications

Most models meet applicable European Community Directives.

Compliance

Rack meets NFPA 1901-12-1.7 mounted in any position.

Warranty and Services

Limited lifetime.

Specifications

Light Source Halogen
Voltage 120 V
Wattage 8 W
Weight (English) 7.4 lb.
Weight (Metric) 3.3 kg
Battery Type 12 AH
Certifications/Compliance NFPA 1901-12-1.7
Color Orange
Description Swivel neck, Standard System
For Use With (Application) Fire and Rescue, Industrial Safety, Utilities, Emergency Medical Services, Homeowners, Sporting Goods, Pest Control
Height (English) 7 in.
Height (Metric) 17.8 cm
Includes Shoulder strap
Length (English) 11.5 in.
Length (Metric) 29.2 cm
Lumen 150 lm
Material ABS Thermoplastic
Product Type Safety Lantern
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.