Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Recirculating Horizontal Electrophoresis Systems PROMO

Catalog No. FBS2025FB25 Shop All Fisherbrand Products
Change view
Click to view available options
Gel Dimensions:
13 x 16 cm
20 x 25 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gel Dimensions
FBS2025FB25 20 x 25 cm
FBS1316FB25 13 x 16 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FBS2025FB25

Choose from four systems.

Minigel System

  • Ideal for quick screening
  • Uses 7 x 10 cm gel tray with two comb positions. Runs up to 16 samples on one gel with combs supplied Can run up to 24 samples on a gel with accessory combs
  • Provides easy, leak-free casting without tape, grease or seals

Midigel Systems

  • Available with proprietary buffer recirculation method which prevents buffer's ionic depletion on long gel runs, keeps ionic gradients from distorting results when multiple sample series are run simultaneously and prevents pH changes that can cause problems in RNA gels
  • Provide leak-free gel casting in 13 x 16 cm gel tray with two comb positions
  • Run up to 32 samples on one gel with combs supplied or up to 48 samples with accessory combs

Large Gel Systems

  • Improve resolution in Northern and Southern blotting of large gels and multiple samples
  • Available with same self-contained buffer recirculation method as Midigel System (FB-SBR-1316)
  • Provide leak-free casting in a 20 x 25 cm gel tray with six comb positions
  • Run up to 36 samples per comb position with supplied combs
  • Permit up to four sets of samples to be run equal distances on the same gel
  • Allow shorter gels to be poured when used with accessory wall comb May also be used with programmable power inverter for field inversion electrophoresis
  • Include a three-point leveling platform

Wide-Format System

  • Permits leak-free casting in an 18 x 23 cm gel tray
  • Runs 25 to 250 samples simultaneously on one gel
  • Produces clear, tight banding patterns with no 'smiling'
  • May be used with included microwell-format combs and a multichannel pipetter to load samples directly from a 96-well plate for faster loading and reduced error
TRUSTED_SUSTAINABILITY

Specifications

Gel Size 20 x 25 cm
Type Horizontal Electrophoresis System
Gel Dimensions 20 x 25 cm
Includes Buffer Chamber, Safety Lid with Attached Power Supply Leads, UV Transparent Gel Tray, Nonskid Rubber Feet, Two 1.5 mm Thick Combs (12 and 20-well)
Model Large Gel System
Sample Size Up to 32 to 48
Volume (Metric) Buffer 2.0 L
Dimensions (L x W x H) 15 x 10.25 x 5.13 in. (38 x 26 x 13 cm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.