Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Recirculation Tube for Dispensette™ S Bottletop Dispensers

Catalog No. 14380881 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Dispensette S, S Organic, S Trace Analysis
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-380-881 Dispensette S, S Organic, S Trace Analysis
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-380-881 Supplier BRANDTECH™ BRAND™ Supplier No. 6747
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Replacement recirculation tube for use with all models of Dispensette™ S.

  • Fits all sizes of Dispensette™ S, Dispensette™ S Organic and Dispensette™ S Trace Analysis bottletop dispensers manufactured in 2016 or later
  • Does not fit Dispensette™ III or Dispensette™ Organic models manufactured prior to 2016

Specifications

Product Type Recirculation Tube
Autoclavable Autoclavable
Color Translucent
Material PTFE
For Use With (Application) Dispensing liquid
For Use With (Equipment) Dispensette S, S Organic, S Trace Analysis
Includes Tube only
Sterility Non-sterile
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.