Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RECK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103291 Supplier Invitrogen™ Supplier No. PA5103291
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RECK.

RECK is a cysteine-rich, extracellular protein with protease inhibitor-like domains whose expression is suppressed strongly in many tumors and cells transformed by various kinds of oncogenes. In normal cells, this membrane-anchored glycoprotein may serve as a negative regulator for matrix metalloproteinase-9, a key enzyme involved in tumor invasion and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RECK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RECK
Gene Accession No. O95980, Q9Z0J1
Gene Alias hRECK; membrane-anchored glycoprotein (metastasis and invasion); membrane-anchored glycoprotein RECK; mRECK; RECK; reversion inducing cysteine rich protein with kazal motifs; reversion-inducing cysteine-rich protein with Kazal motifs; Reversion-inducing cysteine-rich protein with Kazal motifs precursor (mRECK); reversion-inducing-cysteine-rich protein with kazal motifs; ST15; suppression of tumorigenicity 15; suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein with kazal motifs); suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein with kazal motifs); suppressor of tumorigenicity 15 protein
Gene Symbols RECK
Host Species Rabbit
Immunogen A synthesized peptide derived from human RECK(Accession O95980), corresponding to amino acid residues V183-L233.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53614, 8434
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.