Learn More
Abcam Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supplyRabbit monoclonal [EPR18722] to CNOT7Suitable for: Flow Cyt (Intra), ICC/IF, IP, WBKnockout validatedReacts with: Mouse, Rat, Human
Supplier:  Abcam AB195587100UL
Encompass_Preferred
Produced recombinantly (animal-free) for high batch-to-batch consistency and long term security of supplyRabbit monoclonal [EPR18722] to CNOT7Suitable for: Flow Cyt (Intra), ICC/IF, IP, WBKnockout validatedReacts with: Mouse, Rat, Human
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2819151
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.