Learn More
Abcam Recombinant E. coli Maltose Binding Protein, 100UG
Supplier:  Abcam AB167888100UG
Encompass_Preferred
Recombinant E. coli Maltose Binding Protein is a Escherichia coli K-12 Full Length protein, in the 27 to 392 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: b4034, JW3994, malE, Maltose/maltodextrin-binding periplasmic protein, MMBP, Maltodextrin-binding protein, Maltose-binding protein, MBP
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2816685
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.