Learn More
Proteintech Group Inc Recomb. Hmn FGF Basic-TS 100ug
SDP
Supplier:  Proteintech Group Inc HZ1285100UG
Encompass_Preferred
Recombinant human FGFbasic-TS™ is expressed in human 293 cells. FGFbasic is a heparin-binding growth factor and critical for human embryonic stem cells to remain in an undifferentiated state during cell culture. FGFbasic-TS™ is a protein engineered FGFbasic to enchance its thermo- stability without modification of biological function. FGFbasic-TS™ consists of 154 amino acids as a 17 kDa, non-glycosylated monomer.
Catalog No. 50-197-3959
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.