Learn More
Albumin Bioscience Recombinant Human Albumin, Albagen XL in Lyophilized Powder | 1g
Supplier:  Albumin Bioscience 10011G
Encompass
Recombinant Human Albumin, Albagen XL – a high purity, animal free, low endotoxin lyophilized powder. Albagen XL is a patented hypoallergenic recombinant human serum albumin specifically formulated for stability, long-term storage and high monomer content – among the very best in the industry. Chemical properties & structure are equivalent to Human Serum Albumin except for a single deletion at the N-terminus (Asp).
Catalog No. NC1660599
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.