Learn More
Abcam Recombinant human Apolipoprotein E
Supplier:  Abcam AB280330100UG
Encompass_Preferred
Recombinant human Apolipoprotein E (ApoE) is a Human Full Length ApoE protein in the 19 to 317 aa range with >=95% purity, <= 0.005 EU/µg endotoxin level and suitable for SDS-PAGE, mass spectrometry and HPLC. The predicted molecular weight of ab280330 recombinant protein is 34 kDa. - Save time and ensure accurate results - use our recombinant Apolipoprotein E protein (ApoE) as a control - Available in different sizes to fit your experimental needs
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2071566
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.