Learn More
ACROBiosystems Recombinant Protein;Human GITR / TNFRSF18 Protein;HEK293;100UG;GIR,AITR,GITR,TNFRSF18,CD357
Supplier:  ACROBiosystems GIRH5228100UG
Encompass_Preferred
Recombinant Protein;Human GITR / TNFRSF18 Protein;100UG;Human GITR, His Tag (GIR-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Glu 161 (Accession # Q9Y5U5-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.4 kDa. The protein migrates as 22-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GITR, His Tag (Cat. No. GIR-H5228) at 2 μg/mL (100 μL/well) can bind Human GITR Ligand, Fc Tag (Cat. No. GIL-H526a) with a linear range of 1-16 ng/mL (QC tested).
Catalog No. NC0820002
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.