Learn More
Abcam Recombinant Human Angiopoietin 2/ANG2 protein (His tag), 100UG
Supplier:  Abcam AB220589100UG
Encompass_Preferred
Recombinant Human Angiopoietin 2/ANG2 protein (His tag) is a Human Fragment protein, in the 275 to 496 aa range, expressed in HEK 293, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Angiopoietin-2, ANG-2, ANGPT2
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2734337
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.