Learn More
Proteintech Group Inc Recomb. Human TGF beta 2 100ug
SDP
Supplier:  Proteintech Group Inc HZ1092100UG
Encompass_Preferred
Animal-free Recombinant Human TGF beta 2HuXp is expressed from human 293 cells as a mature disulfide-linked homodimer with apparent molecular mass of 25 kDa. This cytokine is produced in a human cell expression system with serum-free, chemically defined media. The cytokine is greater than 95% pure.
Catalog No. 50-197-3881
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.