Learn More
Proteintech Group Inc Recomb. Human TNF alpha 10ug
SDP
Supplier:  Proteintech Group Inc HZ101410UG
Encompass_Preferred
Animal-free Recombinant Human TNF alpha is expressed in human 293 cells and has been shown by gel filtration to be predominantly a non-covalently linked homotrimer, with a molecular mass of 51 kDa. The E. coli form is not a trimer. The cytokine is produced in a human cell express ion system with serum free, chemically defined media.
Catalog No. 50-197-3844
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.