Learn More
MyBioSource RECOMB SALMONELLA TYPHIM 0.1MG
SDP
Supplier:  MyBioSource MBS14695460.1MG
Encompass_Preferred
Salmonella Typhi Outer Membrane Recombinant Protein. Host E-coli / Purity-Purification 95.0 percent as determined by SDS-PAGE. / Form-Format Sterile filtered white lyophilized freeze-dried powder.Lyophilized from 1mg/ml in 20mM sodium carbonate pH-10.For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC3792081