Learn More
Apexbio Technology LLC Recombinant Human BCA-1/CXCL13 500ug
SDP
Supplier:  Apexbio Technology LLC P12150.5
Encompass_Preferred
C-X-C motif chemokine 13 (CXCL13) also known as B-cell-attracting chemokine 1 (BCA-1) is part of the CXC chemokine family encoded by the CXCL13 gene on chromosome 4 It is mainly expressed in secondary lymphoid tissues and guides the migration of B-lymphocytes including B-1 and B-2 subsets through its receptor CXCR5 CXCL13 plays a key role in lymphocyte trafficking follicle development and organization in peripheral lymphoid organs Recombinant human CXCL13 is an 87-amino-acid peptide with 64% sequence identity to murine CXCL13 and 23%-34% homology to other CXC chemokines Recombinant proteins like Human Fibroblast Growth Factor-basic (FGF-basic UniProt P09038 amino acids 1-155) with a His-tag expressed in E coli support studies on the biological functions of these proteins in vitro and in vivo
Catalog No. 50-000-63127
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.