Learn More
Apexbio Technology LLC TNF-alpha/TNFSF2,human recombinant protein 500ug
SDP
Supplier:  Apexbio Technology LLC P12440.5
Encompass_Preferred
Tumor necrosis factor-alpha (TNF- ) a cytokine belonging to the TNF superfamily is primarily produced by activated macrophages neutrophils lymphocytes NK cells endothelial cells and several transformed cell types It exists biologically as both membrane-bound and soluble homotrimeric forms interacting with two distinct receptors (TNFR1 and TNFR2) to mediate apoptotic or proliferative signaling pathways including NF- B and MAPK cascades Due to its central involvement in apoptosis inflammation immune regulation and cell differentiation recombinant human TNF- proteins have extensive applications in biomedical research such as studies on autoimmune disorders inflammation oncology and immunomodulation For instance human TNF- recombinant protein (UniProt P01375 Val77-Leu233) expressed with His-tag in E coli is frequently employed in vitro for investigating cytokine receptor signaling pathways and immune cell-mediated responses
Catalog No. 50-000-64362
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.