Learn More
Apexbio Technology LLC Recombinant Mouse IL-3 500ug
SDP
Supplier:  Apexbio Technology LLC P11620.5
Encompass_Preferred
Interleukin-3 (IL-3) is a cytokine encoded by the IL-3 gene located on chromosome 5 in mice Primarily secreted by activated T cells mast cells and epithelial cells mouse IL-3 plays roles in hematopoiesis by regulating the proliferation differentiation and function of granulocytes and monocyte-macrophage lineage cells IL-3 regulates cellular activities via interaction with the interleukin-3 receptor composed of and subunits initiating downstream intracellular signaling events Mouse IL-3 comprises 140 amino acid residues and exhibits species-specific action due to low homology with its human counterpart Recombinant Mouse IL-3 protein such as Mouse IL-3 (UniProt P01586 Ala27-Cys166) expressed in E coli without tag allows investigation into cytokine-mediated signaling and in vitro hematopoietic assays
Catalog No. 50-000-63409
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.