Learn More
Apexbio Technology LLC GM-CSF, murine recombinant(Synonyms: Granulocyte-Macrophage Colony-Stimulating Factor murine recombinant), 5ug.
SDP
Supplier:  Apexbio Technology LLC P10130.005
Encompass_Preferred
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is a cytokine produced by various cell types including activated T and B lymphocytes, macrophages, mast cells, endothelial cells, and fibroblasts in response to immunologic or inflammatory stimuli. Murine GM-CSF supports proliferation and differentiation of hematopoietic progenitor cells, particularly granulocyte-macrophage precursors, and influences non-hematopoietic cellular functions such as endothelial cell migration and proliferation. Sharing approximately 54% sequence homology with human GM-CSF, murine GM-CSF lacks cross-species biological activity. It is widely utilized in biomedical research models to study hematopoiesis, inflammatory responses, and tumor biology, and holds potential as a therapeutic adjuvant for enhancing immune responses during vaccine development studies.
Catalog No. 50-000-61313
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.