Learn More
Creative Biomart Entolimod, Recombinant agonist of toll-like receptor 5 (TLR5)
Supplier:  Creative Biomart THP0320
Encompass_Preferred
Entolimod is a recombinant protein that acts as an agonist of toll-like receptor 5 (TLR5), an innate immunity receptor. Entolimod activation of TLR5 triggers NF-B signaling, mobilizing an innate immune response that drives expression of numerous genes, including inhibitors of apoptosis, scavengers of reactive oxygen species, and a spectrum of protective or regenerative cytokines.
Catalog No. 50-208-4370
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.