Learn More
Creative Biomart Simoctocog Alfa, Recombinant B-domain deleted (BDD) rFVIII
Supplier:  Creative Biomart THP0092
Encompass_Preferred
The product is a recombinant B-domain deleted (BDD) rFVIII produced in genetically modified human embryonic kidney (HEK) 293F cells. The harvested product is concentrated and purified by a series of chromatography steps. The product is a glycoprotein consisting of 1440 amino acids with an approximate molecular mass of 170 kDa, comprising the FVIII domains A1-A2 + A3-C1-C2 whereas the B-domain, present in the full-length plasma-derived FVIII, has been deleted and replaced by a 16 amino acid linker.
Catalog No. 50-208-4143
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.