Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant B. Thetaiotaomicron Arylsulfatase Protein

Catalog No. 11383SU020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-383-SU020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-383-SU020 Supplier R&D Systems™ Supplier No. 11383SU020
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag

Measured by its ability to cleave 3'-sulfate from Galactose. Recombinant Bt.1636 arylsulfatase will cleave >50% Cy5-Fuc labeled sulfo-G2 (Catalog # GL305), as measured under the described conditions.

Specifications

Accession Number WP_011107907.1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) MolecularWeight-observed: 54-60 kDa, under reducing conditions, MolecularWeight-theroretical: 58 kDa & 59 kDa
Quantity 20 μg
Source E. coli-derived b. thetaiotaomicron Bacteroides Thetaiotaomicron Arylsulfatase protein Met1-Lys509 (S77C) with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Bacteroides Thetaiotaomicron Arylsulfatase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.