Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Bacteria Flagellin NLRC4 Mutant His Protein
SDP

Catalog No. NB171719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171719 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171719 Supplier Novus Biologicals™ Supplier No. NBP318158
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Detected by human & mouse TLR5. Not detected by mouse NLRC4. Activation of TLR5 in human epithelial cell assays based on NF-kB luciferase fusions, in vivo on expression of CCL20 or IL-6. TLR5 can be stimulated in vitro by Flagellin (NLRC4 mutant) (rec.) at 5-50ng/mL & in vivo at a starting concentration of 0.1-1 ug/mouse. Does not activate NLRC4 as tested in bone-marrow mouse macrophages (in the presence of streptolysin O (SLO)). It can be used as negative control for NLRC4 signaling.

Specifications

Accession Number NP_460912
Concentration LYOPH
For Use With (Application) Bioactivity
Formulation Lyophilized from PBS
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.01 EU/μg purified protein (LAL test; Lonza).
Reconstitution Reconstitute with 100 ul sterile water (0.1mg/ml after reconstitution)
Research Category Immunology, Innate Immunity
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Flagellin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.