Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Bacteria NAD Synthetase His Protein
SDP

Catalog No. NB171734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171734 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171734 Supplier Novus Biologicals™ Supplier No. NBP318173
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

∼0.1 U/mg protein (Ammonium or glutamine as nitrogen source) One unit is defined as the amount of enzyme that synthesizes 1umol NAD per min

Specifications

Accession Number P0A5L6
For Use With (Application) Enzyme Activity
Formulation 20mM TRIS-HCl, pH 7.8
Gene ID (Entrez) 55191
Molecular Weight (g/mol) M.W. Theoretical: 75 kDa
Quantity 50 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias EC 6.3.5.1, FLJ10631, FLJ36703, FLJ40627, glutamine-dependent NAD synthetase, glutamine-dependent NAD(+) synthetase, NAD synthetase 1, NAD(+) synthase, NAD(+) synthase [glutamine-hydrolyzing], NAD(+) synthetase
Gene Symbol NADSYN1
Research Category Immunology, Lipid and Metabolism
Purity or Quality Grade >98%, by SDS-PAGE
Protein NAD Synthetase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.