Learn More
Cellular Engineering Technologies Yeast-Expressed Human brain-derived neurotrophic factor (BDNF). Vial of lyophilized power (10µg)
Supplier:  Cellular Engineering Technologies GR100110C
Encompass_Preferred
Human Brain-Derived Neurotrophic Factor (BDNF) is an activity-dependent neurotrophic factor that supports neurons' differentiation, maturation, and survival3 and provides a neuroprotective effect under adverse conditions. BDNF stimulates and controls the growth of new neurons from neural stem cells.
Catalog No. NC2753126