Learn More
Active Motif Recombinant BTK protein
SDP
Supplier:  Active Motif 81083
Encompass_Preferred
BTK (Bruton Tyrosine Kinase), also known as B-Cell Progenitor Kinase, EC 2.7.10.2, AGMX1, ATK, BPK, is a non-receptor tyrosine kinase indispensable for B lymphocyte development, differentiation and signaling. It acts as a platform to bring together a diverse array of signaling proteins and is implicated in cytokine receptor signaling pathways. Binding of antigen to the B-cell antigen receptor (BCR) triggers signaling that ultimately leads to B-cell activation. After BCR engagement and activation at the plasma membrane, BTK can phosphorylate PLCG2 at several sites, igniting the downstream signaling pathway through calcium mobilization, followed by activation of the protein kinase C (PKC) family members. It also plays an important role in the function of immune cells of […] For more information, please visit: https://www.activemotif.com/catalog/details/81083.
Catalog No. NC1851443
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.