Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant C. difficile Toxin A/TcdA Protein

Catalog No. 8619GT020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8619GT020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8619GT020 Supplier R&D Systems™ Supplier No. 8619GT020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to hydrolyze UDP-Glucose. The specific activity is >15 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P25738 (msyB) & P16154 (TcdA)
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Molecular Weight (g/mol) M.W. (Observed): 78 kDa, reducing conditions; M.W. (theoretical): 79 kDa
Quantity 20 μg
Source E. coli-derived c. difficile Toxin A/TcdA protein msyB (Thr2-Arg124) (N-terminus) Accession # P25738 GSENLYFQGH C. difficile TcdA (Met1-Leu542, Arg184Asn) (N-terminus) Accession # P16154 HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias tcdA, ToxA, Toxin A
Common Name Clostridium Difficile Toxin A
Gene Symbol tcdA
Biological Activity Measured by its ability to hydrolyze UDP-Glucose. The specific activity is >15 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species C. difficile
Recombinant Recombinant
Expression System E. coli-derived msyB (Thr2-Arg124) Accession # P25738, N-terminus; GSENLYFQGH; C. difficile TcdA (Met1-Leu542, Arg184Asn) Accession # P16154; HHHHHH, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Clostridium Difficile Toxin A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.