Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Canine FGF basic/FGF2/bFGF Protein

Catalog No. 8575FB025CF
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8575FB025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8575FB025CF Supplier R&D Systems™ Supplier No. 8575FB025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Canine FGF basic/FGF2 Protein is derived from E. coli. The Recombinant Canine FGF basic/FGF2 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number FIP6K6
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 TCEP and EDTA.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) Observed M.W.: 17kDa, reducing conditons, Theoretical M.W.: 16kDa
Quantity 25 μg
Source E. coli-derived canine FGF basic/FGF2/bFGF protein Pro10-Ser155 (Lys36Asn)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Common Name FGF basic/FGF2
Gene Symbol FGF2
Biological Activity Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is typically 0.2-1ng/mL.
Conjugate Unconjugated
Species Canine
Recombinant Recombinant
Expression System E. coli-derived Pro10-Ser155 (Lys36Asn)
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.