Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 403977 |
| Molecular Weight (g/mol) | 36.3 kDa (p40), 23.6 kDa (p35) |
| Name | IL-12 |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived canine IL-12 protein, (N-terminus) Canine IL-12 p40 (Ile23-Ser329) Accession &Num; Q28268 10-His tag Canine IL-12 p35 (Arg26-Ser222) (Asp63Glu,Tyr64Leu,Ile65Tyr,Pro66Ser) Accession &Num; Q28267 10-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
