Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Chicken Netrin-1 Protein
SDP

Catalog No. 128N1025CF Shop All R&D Systems Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
128N1025CF Lyophilized from a 0.2μm filtered solution in PBS.
128N1025 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 128N1025CF Supplier R&D Systems™ Supplier No. 128N1025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Chicken Netrin-1 Protein is derived from NS0. The Recombinant Chicken Netrin-1 Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 396389
Molecular Weight (g/mol) 66.3 kDa
Name Netrin-1
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived chicken Netrin-1 protein, (N-terminus) Chicken Netrin-1 (Gly26-Ala606) Accession &Num; Q90922 DIEGRGGGSGGGSGGGS 10-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.