Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cotton Rat CD8 alpha Protein

Catalog No. 9648CD050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9648CD050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9648CD050 Supplier R&D Systems™ Supplier No. 9648CD050
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Cotton Rat CD8 alpha Protein CF is a dimeric complex made up of two Ig superfamily members.

The Recombinant Cotton Rat CD8 alpha Protein is derived from NS0.

Specifications

Accession Number AAL55392
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 925
Molecular Weight (g/mol) 18 kDa
Name CD8, CD8A, CD8a molecule, LEU2, p32
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived cotton rat CD8 protein Ser24-Asp182,with a C-terninal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Cotton Rat
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.