Learn More
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS, NaCl and Sucrose. |
| Molecular Weight (g/mol) | 17 kDa |
| Name | IFN-gamma |
| Quantity | 25 μg |
| Source | E. coli-derived cotton rat IFN-gamma protein His21-Ile170,with an N-terminal Met and two substitutions,G150R,Y165N Accession &Num; Q9QXX2 |
| Storage Requirements | 3 months from date of receipt, 2 to 8° C as supplied. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
