Learn More
R&D Systems™ Recombinant Cyno/Rhesus Lysyl Oxidase Homolog 2 Protein, CF
Description
Specifications
Specifications
| Accession Number | XP_005562900.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 4017 |
| Molecular Weight (g/mol) | M.W. (Observed): 92-102 kDa, under reducing conditions, M.W. (Theroretical): 85 kD |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived Lysyl Oxidase Homolog 2/LOXL2 protein Gln26-Gln774, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.