Learn More
R&D Systems™ Recombinant Cyno u-Plasminogen Activator (uPA) Protein
Description
Specifications
Specifications
| Accession Number | XP_05565501.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl and CaCl2. |
| Gene ID (Entrez) | 5328 |
| Molecular Weight (g/mol) | M.W.-observed: 32-41 kDa (short form) and 48-59 kDa (long form), non-reducing conditions, M.W.-theroretical: 18 kDa (long A chain), 3 kDa (short A chain), 30 kDa (B chain) |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey u-Plasminogen Activator (uPA)/Urokinase protein Ser21-Leu430 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.