Learn More
R&D Systems™ Recombinant Cynomolgus 4-1BB Ligand/TNFSF9 His Protein
Specifications
Specifications
| Accession Number | XP_015296398.1 |
| Formulation | Lyophilized from a 0.2 μm filtered solution in HEPES, NaCl and TCEP with Trehalose. |
| Gene ID (Entrez) | 8744 |
| Molecular Weight (g/mol) | M.W. Observed: 19-23 kDa, under reducing conditions, M.W. Theoretical: 20 kDa |
| Quantity | 100 μg |
| Source | E. coli-derived cynomolgus monkey 4-1 BB Ligand/TNFSF9 protein Arg68-Glu251, with an N-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | 4-1 BB ligand, 41 BB Ligand, 4-1 BBL, 4-1 BB-L, CD137L, homolog of mouse 4-1 BB-L, receptor 4-1 BB ligand, TNFSF9, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.