Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus 5'-Nucleotidase/CD73 His Protein

Catalog No. 10173EN025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-173-EN025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-173-EN025 Supplier R&D Systems™ Supplier No. 10173EN025
Add to Cart
Add to Cart
This item is not returnable. View return policy

CD73, known as ecto-5'-Nucleotidase, converts extracellular nucleoside 5' monophosphates to nucleosides, with AMP as its preferred substrate

Measured by its ability to hydrolyze the 5’-phosphate group from the substrate adenosine-5’-monophosphate (AMP). The orthophosphate product is measured by a Malachite Green Phosphate Detection Kit (Catalog # DY996 ). The specific activity is >6500 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number XP_005552488.1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2 and Glycerol.
Gene ID (Entrez) 4907
Molecular Weight (g/mol) M.W. (Observed): 57-75 kDa; M.W. (theoretical): 59 kDa
Purification Method >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey 5 '-Nucleotidase/CD73 protein Trp27-Lys547 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Buffer Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2 and Glycerol
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein 5 '-Nucleotidase/CD73
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.