Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Cynomolgus B7-H6 Fc Chimera Protein

Catalog No. 8984B701M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-84B-701M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-84B-701M Supplier R&D Systems™ Supplier No. 8984B701M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to induce IFN-gamma secretion by NK-92 human natural killer lymphoma cells. Brandt, C.S. et al. (2009) J. Exp. Med. 206:1495. The ED50 for this effect is 0.5-2.5 μg/mL.

Specifications

Accession Number XP_005578557
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 374383
Molecular Weight (g/mol) M.W. (Observed): 70-81 kDa, reducing conditions; M.W. (theoretical): 53 kDa
Quantity 1 mg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey B7-H6 protein Cynomolgus B7-H6 (Asp25-Ser262) (N-terminus) Accession # XP_005578557 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias B7 homolog 6, B7H6, B7-H6, DKFZp686I21167, DKFZp686O24166, NCR3LG1
Gene Symbol NCR3LG1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.